Cole-Parmer High-Temp Extended-Length Conductivity Cell, K=0.1, RTD
Representative image only.
Cole-Parmer Item # WW-19501-77
Rugged sensors withstand high pressure and temperature
  • Designed for in-line or submersion conductivity measurements in processes that generate up to 250 psi and 392°F (200°C)
  • 316 stainless steel construction provides excellent chemical compatibility
  • Available with automatic temperature compensation (ATC) for compatibility with most makes and models of conductivity controllers
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Cell ConstantK = 0.1
  • Cell TypeContact
  • Electrode Material316 SS
  • Body Material316 SS
  • Fitting¾\" NPT(M)
  • Mounting TypeThreaded
  • Max temperature (° C)200
  • Max temperature (° F)392
  • Length (cm)7.6
  • Length (in)3
  • Cable Length (m)0.15
  • Cable Length (ft)0.5
  • DescriptionHigh-Temp Extended-Length Conductivity Cell, K=0.1, RTD
  • Warranty1 year
1 Year Warranty
Key Features
  • Designed for in-line or submersion conductivity measurements in processes that generate up to 250 psi and 392°F (200°C)
  • 316 stainless steel construction provides excellent chemical compatibility
  • Available with automatic temperature compensation (ATC) for compatibility with most makes and models of conductivity controllers
  • 1 year warranty
More About this Item
Cole-Parmer industrial conductivity cells are designed for high performance in harsh measurement conditions. Stainless steel construction resists aggressive chemicals, high pressures, and high temperatures. Threaded ¾” NPT electrode bodies allow for in-line or submersion mounting. Cells are well-suited for boiler or ultrapure water applications.