Cole-Parmer Heat Exchanger for Fermentation/Bioreactor System with 18-mm Size Port, 2-5 L
Representative image only.
Cole-Parmer Item # WW-01919-80
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithCole-Parmer Modular Fermentation Systems
  • DescriptionHeat Exchanger for Fermentation/Bioreactor System with 18-mm Size Port, 2-5 L
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Heat exchangers circulate warm or cold water to maintain vessel tempperature. Not recommended for cell culture systems.