Cole-Parmer Essentials Handheld Tally Counter, Stainless Steel, 4-Digit
Representative image only.
Cole-Parmer Essentials Item # WW-98766-27
Counting made easy
  • Numbers instantly and automatically return to zero with a twist of the reset knob
  • Protective window seals out dust, dirt and humidity
  • Corrosion-proof stainless steel outer material for long-life
PRODUCT OPTIONS
₹2,500.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Width (in)1 1/8
  • Colony Counter TypeMechanical Handheld
  • Display4-Digit
  • MaterialStainless Steel
  • Diameter (in)1.75
  • DescriptionHandheld Tally Counter, Stainless Steel, 4-Digit
1 Year Warranty
Includes
Loop.
Key Features
  • Numbers instantly and automatically return to zero with a twist of the reset knob
  • Protective window seals out dust, dirt and humidity
  • Corrosion-proof stainless steel outer material for long-life
More About this Item
The laboratory standard tool for counting bacterial colonies or tallying any repetitive event. Four-digit counter features a quick reset knob and a lens that makes the digits very easy to read. No-fail mechanism and tough, outer corrosion-proof stainless steel case ensure years of use.
Handheld counter has a finger ring for a secure, comfortable grip.