Cole-Parmer Graphite Straight Ferrules for Packed Columns, 1/4"; 10/Pk
Representative image only.
Cole-Parmer Item # WW-01992-02
Reusable one-piece design
  • Constructed of durable graphite
PRODUCT OPTIONS
₹5,000.00INR / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionGraphite straight ferrules, 1/4\"; 10/pack
Key Features
  • Constructed of durable graphite
More About this Item
Graphite Ferrules are preferred for capillary columns. Maximum temperature: 450degC. Does not stick to glass and is reusable if not over-tightened. One-piece design – no back ferrule.