Cole-Parmer Grade 54 Ashless Filter Paper, 18.5 cm Diameter; 100/Pk
Representative image only.
Cole-Parmer Item # WW-06648-49
Perfect for gravimetric analysis or sample preparation
  • High-purity, acid-washed filters with high wet strength
  • 6 month warranty
PRODUCT OPTIONS
₹9,700.00INR / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Pore Size (micron)10
  • Diameter (mm)185
  • Diameter (cm)18.5
  • DescriptionGrade 54
  • Warranty6 month
6 Month WarrantyMade in the U.S.A.
Key Features
  • High-purity, acid-washed filters with high wet strength
  • 6 month warranty
More About this Item
Grade 94 filters are fine retention filters for analysis of fine particles such as barium sulfates, lead sulfates, stannic or nickel sulfide, calcium oxalate, and calcium fluoride. Grade 74 filters are well-suited for general filtration such as analysis of clay, copper, bismuth, silicon, and iron. Grade 54 filters are ideal for filtration of coarse or gelatinous precipitate, namely iron, zirconium, or aluminum hydroxides, and cobalt sulfide. Sold 100/pk.