Cole-Parmer Glassware Washing Kit; Size 10 Gloves, 10 qt Bin, Alconox
Representative image only.
Cole-Parmer Item # WW-25000-08
Bundled kits for added convenience and to save you money!
  • Choose from the glove size, bin size, and cleaner that best suits your needs
SPECS & DESCRIPTION
Specifications & Description
  • FormatPowder
  • ApplicationsManual or ultrasonic cleaning
  • DescriptionGlassware Washing Kit; Size 10 Gloves, 10 qt Bin, Alconox
Includes
Key Features
  • Choose from the glove size, bin size, and cleaner that best suits your needs
More About this Item
All kits include two types of brushes and cleaning gloves. Use the wide, 15-1/2" long brush for larger glassware like bottles, beakers, and flasks; and one of the twelve tapered, 17" long brushes for smaller and thin-necked glassware like test tubes, pipettes, cuvettes, and burettes. Twelve pairs of reusable cleaning gloves are right- or left-handed, unlined for easy on/off, and made of nitrile for excellent chemical resistance and durability. The 13 inch long length helps provide additional protection.