Cole-Parmer Glass Filtration Assembly, 47 mm Diameter, 300 mL; Stopper with 9/16" Diameter Hole
Representative image only.
Cole-Parmer Item # WW-06644-75
Ideal for filtering small to medium volumes of liquid
  • Sintered glass support provides an even, flat support structure for membrane filters
  • Minimize contact between sample and filtration system
  • Complete filtration assembly — just add a membrane
  • Silicone rubber stopper features 3/8" diameter hole
  • 1 year warranty
PRODUCT OPTIONS
₹23,300.00INR / Set of 1
SPECS & DESCRIPTION
Specifications & Description
  • Filter Diameter (mm)47
  • Body MaterialGlass
  • Funnel Size (mL)300
  • SupportSintered Glass
  • Stopper Size8
  • DescriptionGlass Filtration Assembly, 47 mm Diameter, 300 mL; Stopper with 9/16\" Diameter Hole
  • Warranty1 year
1 Year Warranty
Includes
300 mL borosilicate glass funnel and base, no. 8 silicone rubber stopper with hole, and anodized aluminum clamp.
Key Features
  • Sintered glass support provides an even, flat support structure for membrane filters
  • Minimize contact between sample and filtration system
  • Complete filtration assembly — just add a membrane
  • Silicone rubber stopper features 3/8" diameter hole
  • 1 year warranty
More About this Item
These Cole-Parmer glass filtration assemblies will accept a 47-mm membrane and will fit perfectly into a filter flask with no. 8 stopper neck size. Glass filter holder assembly consists of a 300-mL borosilicate glass funnel and base, no. 8 silicone rubber stopper with hole, and anodized aluminum clamp.
These filtration assemblies are designed for HPLC, particulate, and microbiological contamination filtrations of particulate analysis.