Cole-Parmer Gel Tray for Horizontal Mid-Size Gel System Model 28560-10
Representative image only.
Cole-Parmer Item # WW-28560-72
For Gel system 28560-00
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionGel Tray for Horizontal Mid-Size Gel System Model 28560-10
  • Warranty3 year
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
Gel Trays let you pour gels in the unit using the tapered baffles; outside of the unit by taping the ends of the tray; or outside of the unit using the gel casters sold seperately. Trays are UV-transparent acrylic. Multiple comb slots let you cast gels for simultaneous short runs.
With 4 comb slots; pathlength of 3.1 cm/slot