Cole-Parmer Fluorescent Flt Yellowith Green Dye Powder, 1 Lb
Representative image only.
Cole-Parmer Item # WW-00298-27
Determine water flow in closed circuits or open channels
  • Biodegradable and nontoxic
PRODUCT OPTIONS
₹32,800.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Reagent PlatformPowder
  • Gallons Of Water For 1 ppm (Gal)120000
  • DescriptionFluorescent Flt Yellowith Green Dye Powder, 1 Lb
Key Features
  • Biodegradable and nontoxic
More About this Item
Dyes diffuse rapidly in water, producing a vivid, easy-to-see color. Excellent for identifying diffusion and dispersion patterns, rates of flow, discharge locations, seepage areas, sea marking, and more.
Fluorescent yellow/green dye (fluorescein) works best in silt-filled waters. Use an ultraviolet lamp (order separately below) to trace fluorescent red or yellow/green dyes where normal visual detection is difficult. Minimum detectable concentration of all dyes in clear water is 1 ppm.