Cole-Parmer Essentials Solid Black Rubber Stoppers, Standard Size 8.5; 12/Pk
Representative image only.
Cole-Parmer Essentials Item # WW-62990-28
SPECS & DESCRIPTION
Specifications & Description
  • Standard Size Number8.5
  • Top Outside Diameter (OD) (mm)43
  • Bottom Outside Diameter (OD) (mm)36
  • Height (mm)25
  • ColorBlack
  • MaterialRubber
  • DescriptionSolid Black Rubber Stoppers, Standard Size 8.5; 12/Pk
More About this Item
Black rubber stoppers stay pliant over a long period and withstand laboratory conditions without hardening. Stoppers retain resiliency even after repeated autoclaving. Each stopper has its size in raised figures on top. Contain some sulfur. Autoclavable.