Cole-Parmer Essentials Single-Ply Cellulose Critical Task Wipes and Wipe Dispenser Bundle
Representative image only.
Cole-Parmer Essentials Item # WW-78902-94
Eco-friendly lab wipes made from 100% recycled cellulose
  • Ideal for optics and other sensitive surfaces
  • Perfect wipe for light-duty tasks and delicate surfaces
PRODUCT OPTIONS
₹6,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Material100% recycled cellulose
  • Width (in)4 2/5
  • Length (in)8 19/64
  • DescriptionSingle-Ply Cellulose Critical Task Wipes and Wipe Dispenser Bundle
Includes
Key Features
  • Ideal for optics and other sensitive surfaces
  • Perfect wipe for light-duty tasks and delicate surfaces
More About this Item
Made from 100% recycled cellulose (50% post consumer), these environmentally friendly lab wipes are excellent for your light-duty applications, will not scratch, and can be used in a variety of industries. The wipes are made of single-ply, absorbent, and soft material making them ideal for cleaning small parts, lenses, and other equipment in your lab facility. Since they are low linting and low in chemical extractables, they’ll leave surfaces extremely clean.
Packaged in recycled and biodegradable boxes, the wipes are a complete ecologically friendly wipe option. The 4.4″ x 8.3″ (11 x 21 cm) wipes are packaged 280 per box.