This item is now discontinued. Please browse similar items
Cole-Parmer Essentials LiteSafe® 50 mL Black LiteSafe Centrifuge Tube - 50/pk
Representative image only.
Cole-Parmer Essentials Item # WW-06344-35
Protect photosensitive samples
  • Unlike amber tintted tubes, these are completely opaque, protecting even the most light sensitive samples
  • DNase, RNase, and Pyrogen free
  • Ideal for use with fluorescent probes or ethidium bromide
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • Max RCF x g10000
  • AutoclavableYes
  • Tube Capacity (mL)50
  • Tube MaterialPolypropylene
  • Tube ColorBlack
  • Tube Bottom ShapeConical
  • Cap IncludedYes
  • Cap StyleScrew
  • GraduationsNone
  • Description50 mL Black LiteSafe Centrifuge Tube - 50/pk
Key Features
  • Unlike amber tintted tubes, these are completely opaque, protecting even the most light sensitive samples
  • DNase, RNase, and Pyrogen free
  • Ideal for use with fluorescent probes or ethidium bromide
More About this Item
Disposable polypropylene tubes and screw caps that have recessed HDPE lids to prevent leaking. Tubes feature a frosted writing surface for permanent identification and labeling. Autoclavable to 122° C and can be stored down to -100° C.