Cole-Parmer Essentials Indirect Vent Autoclavable Goggles with Sealed Bottom; 10 pcs/box, 10 boxes/case

Representative image only.
Cole-Parmer Essentials Item # WW-16392-28
Prevent contamination entering cleanroom
  • Tested to withstand 35 autoclave cycles at 121°C, 30 minutes
  • No deformation or change in transparency due to autoclaving
  • Soft thermoplastic rubber frame provides utmost comfort
  • Fit over most prescription glasses
  • 3 year warranty

PRODUCT OPTIONS

₹157,000.00INR / Box of 100

SPECS & DESCRIPTION

Specifications & Description

  • Frame ColorBlue
  • ComplianceEN166:2001, ANSI Z87.1:2015
  • DescriptionIndirect Vent Autoclavable Goggles with Sealed Bottom; 10 pcs/box, 10 boxes/case
  • Warranty3 years
3 Year Warranty

Key Features

  • Tested to withstand 35 autoclave cycles at 121°C, 30 minutes
  • No deformation or change in transparency due to autoclaving
  • Soft thermoplastic rubber frame provides utmost comfort
  • Fit over most prescription glasses
  • 3 year warranty

More About this Item

Sterilization:
• Tested to withstand 35 autoclave cycles at 121°C, 30 minutes
• No deformation or change in transparency, due to autoclaving

Ventilation:
• Indirect vented goggles

Ergonomics:
• Thermoplastic rubber Frame provides utmost comfort
• 24mm Non-irritant Silicone strap provides easy cleaning
• Light in weight : 99.99 gms

User Comfort:
• Wide vision goggle provides high level of comfort & maximum protection
• Scratch-resistant polycarbonate lens with Premium anti-fog coating
• Triple-hollows system facilitate strap length adjustment
• Fit over most prescription glasses

Certification:
• EN166:2001
• ANSI Z87.1:2015

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat