Cole-Parmer Electronic Pipettor, Single Channel; 0.5 to 10 uL
Representative image only.
Cole-Parmer Item # WW-16344-66
Three-point auto-calibration provides uncompromised precision
  • Graphical interface reduces the learning curve and makes it extremely easy to use
  • Extremely lightweight-for effortless and pain-free pipetting even after extensive use
  • Stores and recalls the three most frequently used modes
  • Intelligent sleep mode enhances battery run-time by more than 40%
  • Efficient battery delivers 5 hours of continuous use after a 2-hour charge
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • AutoclavableYes-tip cone
  • Max Volume (µL)10
  • Min Volume (µL)0.5
  • Accuracy (%)±2.5
  • Number Of Channels1
  • Tip EjectorYes
  • Auto ShutoffYes
  • Calibration3 point auto-calibration
  • BatteryLithium ion
  • DescriptionElectronic Micropipette, Single Channel; 0.5 to 10 uL
  • Warranty2 year
Two year warrantyConformite EuropeenneNIST Traceable Calibration
Key Features
  • Graphical interface reduces the learning curve and makes it extremely easy to use
  • Extremely lightweight-for effortless and pain-free pipetting even after extensive use
  • Stores and recalls the three most frequently used modes
  • Intelligent sleep mode enhances battery run-time by more than 40%
  • Efficient battery delivers 5 hours of continuous use after a 2-hour charge
  • 2 year warranty
More About this Item
Cole-Parmer electronic pipettor has been developed for a truly effortless and intuitive pipetting experience. Its unique ergonomic design allows for hours of continuous pipetting without any pain. Five different pipetting modes include forward pipetting, reverse pipetting, multi-dispensing, dilution, and mixing. Flexible charging—via the USB charger or optional charging stand for overnight charging.