Cole-Parmer Electronic Burette, Bottletop Dispenser, 10 mL, 230V, 50Hz

Representative image only.

Cole-Parmer Electronic Burette, Bottletop Dispenser, 10 mL, 230V, 50Hz

Cole-Parmer | Item: 1597789

Get better accuracy with motor-controlled filling and dispensing

₹239,000.00INR / Each

₹239,000.00INR / Each
Product Description

Includes

E-Burette, controller, amber window, telescopic tube, adapter set (28, 32, 36, 40, 45 mm), USB drive for software and driver, mini-USB cable, stylus, charging AC adaptor, and 2000 mAh Li-ion rechargeable batteries, and calibration certificate.

Key Features

  • Easy, user-friendly, TFT touch screen helps to perform quick and accurate titrations
  • Motor-controlled piston movement, help in eliminating the errors
  • Three pre-set dispensing speeds allows achieve the end point very accurately.
  • Conforms to ISO 8655 standards & GLP compliant.
  • 1 year warranty

More About this Item

Accuracy of Cole-Parmer E-Burette is ± 0.2% and able to perform precise titrations within the error limits of 'Class A' glass burettes. 20 results can be saved in the memory. Computer connectivity is available through a micro-USB cable for transferring and recording data. Rechargeable, long-life, replaceable batteries are provided in the control panel. Amber colored window is provided as an accessory for those chemicals/reagents that are sensitive to light

1 Year WarrantyNIST Traceable Calibration
Specifications
  • Capacity (mL)10
  • Power (VAC)230
  • DescriptionElectronic Burette, Bottletop Dispenser, 10 mL, 230V, 50Hz
  • Warranty1 year
Reviews
Q&A