Cole-Parmer Electrical Tape, 7 mil, 3/4" x 66'. 66, Black
Representative image only.
Cole-Parmer Item # WW-08277-07
Soft PVC backing with flame-retardant rubber-based adhesive
SPECS & DESCRIPTION
Specifications & Description
  • ColorBlack
  • Thickness (mil)7
  • Width (in)3/4
  • Length (ft)66
  • Width (cm)1.9
  • Length (m)20.12
  • DescriptionElectrical Tape, 7 mil, 3/4\" x 66'. 66, Black
More About this Item
Excellent all weather tape with temperature resistance from 14 to 176°F (-10 to 80°C). Used for general insulating and protection of electrical wires.