Cole-Parmer Eight-element probe for 130-Watt ultrasonic processors
Representative image only.
Cole-Parmer Item # WW-04712-08
  • 3 year warranty
₹216,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionEight-element probe for 130-Watt ultrasonic processors
  • Warranty3 year
3 Year WarrantyConformite EuropeenneISO9001:2000 Certified Supplier
Key Features
  • 3 year warranty
More About this Item
This eight-element probe provides cost-effective processing of 250 µL to 2 mL samples simultaneously. The energy delivered to each tip is identical and approximately the wattage delivered by the power supply divided by the number of microtips. The probe consists of an aluminum coupler and eight microtips spaced 23/64" (9 mm) apart. Order power supply separately.