Cole-Parmer DX53056101 Stereozoom Microscope, built-in 2.0 mega pixel digital camera, 110-220 VAC
Representative image only.
Cole-Parmer Item # WW-48925-40
Large stage allows easy positioning of larger objects
  • Software enables you to count and measure objects
  • Add audio comments to saved images
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Magnification10x to 40x
  • Maximum Magnification40x
  • Head ConfigurationBinocular
  • Resolution1600 x 1280
  • Objectives1x to 4x
  • Eyepiece Magnification10x
  • EyepiecesWidefield
  • Stage Dimensions80 mm diameter
  • Illuminator(s)Halogen with rheostat, 20 W
  • Stand TypeStandard
  • Image Sensor1/2\" CMOS
  • Power (VAC)110 / 220
  • DescriptionStereozoom Microscope, built-in 2.0 mega pixel digital camera, 110-220 VAC
  • Warranty1 Year
1 Year WarrantyConformite EuropeenneUL Listed.
Key Features
  • Software enables you to count and measure objects
  • Add audio comments to saved images
  • 1 year warranty
More About this Item
A simple USB connection with your PC or Mac lets you put this plug-and-play stereozoom microscope to work. The built-in digital camera features powerful analysis and manipulation software that counts and measures objects. Measure length in metric or English scales, ellipses, rectangles, and irregular shapes. Compute areas of an entire image or portions of it. Combine images, each with a different focus, into one clear image. Enhance saved images with audio narration recordings using MIG format. Software requires a minimum of Windows® 98 or Mac OS® 9.01, 64 MB RAM, and a 4 MB display card (not included).
Zoom continuously through a range of 1x to 4x with incident and/or transmitted illumination—rheostat light control ensures clean image views, and the auto white balance function corrects the color. This stereozoom microscope has widefield 10x eyepieces with dual diopter and interpupillary adjustability from 54 to 76 mm.