Cole-Parmer Digital Round Top Ceramic-Coated Stirring Hot Plate with Counter Reaction
Representative image only.
Cole-Parmer Item # WW-15956-32
Ceramic-coated top plate combines temperature uniformity with durability
  • Aluminum plate ensures optimal heat transfer while ceramic coating promotes excellent chemical resistance
  • Counter-reaction maintains a constant speed, even with viscosity changes
  • 3 year warranty
₹55,900.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Top Plate MaterialCeramic-coated Aluminum
  • Min Temperature (° F)75
  • Max Temperature (° F)698
  • Min Temperature (° C)20
  • Max Temperature (° C)370
  • No. Of Stirring Positions1
  • DescriptionDigital Round Top Ceramic-Coated Stirring Hot Plate with Counter Reaction
  • Warranty3 Years
3 Year Warranty
Key Features
  • Aluminum plate ensures optimal heat transfer while ceramic coating promotes excellent chemical resistance
  • Counter-reaction maintains a constant speed, even with viscosity changes
  • 3 year warranty
More About this Item
This digital hot plate stirrer ensures precise thermoregulation of the plate as well as a high degree of reliability and safety. The control panel is separated from the heating plate, and leaks or spills are channelled away from the controls by the dedicated run-off groove. The pressure die-cast structure ensures that accidental liquid spills cannot reach the internal parts of the unit. The raised front panel features easy-to-use temperature and speed controls and the ceramic-coated heating plate provides easy cleaning and resistance to scratches and surface abrasions.