Sherwood Scientific Digital Interface/Linearizer Module for Single-Channel Flame Photometers
Representative image only.
Sherwood Scientific Mfr # 41086001 – Item # WW-02656-10
Easily connects to a printer or computer
  • Mounts directly to front cover of flame photometer
  • Ability to linearize sodium curves
  • 1 year warranty
₹277,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDigital Interface/Linearizer Module for Single-Channel Flame Photometers
  • Warranty1 year
1 Year Warranty
Key Features
  • Mounts directly to front cover of flame photometer
  • Ability to linearize sodium curves
  • 1 year warranty
More About this Item
The digital interface allows you to connect the single-channel flame photometer to a printer or a computer. Print results with sequence number, designation (blank, calibration, sample, and repeat), and date/time stamp. Mounts directly onto the front cover of the flame photometer. The module also features the ability to linearize sodium curves for measuring in the 100 to 200 mmol/liter range typical of serum samples.