Cole-Parmer DIGITAL CERAMIC STIRRING HOT PLATE WITH COUNTER REACTION
Representative image only.
Cole-Parmer Item # WW-15956-31
Counter-reaction maintains a constant speed, even with viscosity changes
  • Top plate and body resist acids, bases, and solvents
  • Controls and top plate are separated to increase safety
  • Digital “HOT PLATE” warning signal when the plate temperature is above 50°C
  • 2 year warranty
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Top Plate MaterialCeramic
  • No. Of Stirring Positions1
  • DescriptionDIGITAL CERAMIC STIRRING HOT PLATE WITH COUNTER REACTION
  • Warranty2 Years
Two year warranty
Key Features
  • Top plate and body resist acids, bases, and solvents
  • Controls and top plate are separated to increase safety
  • Digital “HOT PLATE” warning signal when the plate temperature is above 50°C
  • 2 year warranty
More About this Item
Easy-to-clean, white ceramic top is highly resistant to corrosion and is excellent for observing liquid colour changes. Built-in electronics keep stirring speeds constant during heavy loads or changing viscosities. Microprocessor control allows for precise temperature and speed control. Control panel is separated from the main body, protecting electronics from splashes and spills. Stirrer features a "HOT PLATE" warning signal that remains illuminated while the temperature is on and will flash if the instrument is turned off until plate temperature drops below 50°C.