Includes four different power receptacle adapters to convert 115/220 VAC to 9 VDC
Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
Easily keeps track of results for thermometers, hygrometers, barometers, light meters, anemometers, manometers, dissolved oxygen, conductivity and more
Software on CD and 6-foot cables (USB, serial, and instrument connections)
Key Features
Includes four different power receptacle adapters to convert 115/220 VAC to 9 VDC
Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
Easily keeps track of results for thermometers, hygrometers, barometers, light meters, anemometers, manometers, dissolved oxygen, conductivity and more
1 year warranty
More About this Item
This VAC/VDC adapter features four power receptacle/plug adapters.
Allows up to eight unique Traceable® meters to simultaneously capture data and send it to directly to email to any smartphone or computer. Readings are saved to a file that can be viewed, printed as-is, or merged in a report or spreadsheet format. Data continues to be collected in background even when user works on other software programs. 24/7 audible/visual alarm reminds user to check on data. Powerful and easy to use computer data capture program works with Traceable® Instruments with computer output. Records interval readings from 1 to 10,000 seconds; displays minimum/maximum readings; and utilizes an alarm mode that permits the user to be notified visually, audibly, and by email when an alarm is triggered. Networking server/client capability allows the captured data to be monitored on a remote workstation. Readings may be streamed to cell phones or anywhere via email every minute or in any time interval. 24/7 notification of audible/visual alarms are sent to user’s computer, remote computer, via email anywhere, and by email to cell phones. Program automatically installs in less than two minutes. Absolutely no user programming, no user keystrokes, no user entries are required. Simply connect the cable to the instrument and to a USB port (or serial port) and data is captured. Operation is straight forward plug and play. It is that easy. It is designed to work with Windows® 98/Me/NT/2000/XP/Vista/Windows 7/Windows 8/Windows 10.