Cole-Parmer Comb for Vertical Slab Gel System; 14 Wells, 0.75 mm Thick
Representative image only.
Cole-Parmer Item # WW-28572-52
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Comb Thickness (mm)0.75
  • Number of Wells14
  • DescriptionComb for Vertical Slab Gel System; 14 Wells, 0.75 mm Thick
  • Warranty3 year
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
Combs of PTFE fluoropolymer are precision machined to ensure consistent well width and depth. Combs are easy to remove from gels — leaving smooth, clean sample wells.