Cole-Parmer Comb for Horizontal Mid-Size Gel System; 20 Wells, 1.0 mm Thick
Representative image only.
Cole-Parmer Item # WW-28560-53
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Comb Thickness (mm)1
  • Number of Wells20
  • DescriptionComb for Horizontal Mid-Size Gel System; 20 Wells, 1.0 mm Thick
  • Warranty3 year
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
Combs are single sided and height adjustable; available in three thicknesses to accommodate different sample volumes. The 13- and 27- well combs are spaced to allow sample loading with multi-channel pipettors.