Cole-Parmer COElectrode with 12-mm body diameter "H" length: 300 mm
Representative image only.
Cole-Parmer Item # WW-29000-05
Measure carbon dioxide
  • In-situ-sterilizable
  • Ideal even for small-volume fermentors
  • Special internal pH electrode lets you calibrate without using gases
  • 1 year warranty
PRODUCT OPTIONS
₹167,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Meter TypeAccessory; Probes
  • Length (cm)30
  • Diameter (cm)1.2
  • DescriptionCOElectrode with 12-mm body diameter \"H\" length: 300 mm
  • Warranty1 year
1 Year Warranty
Key Features
  • In-situ-sterilizable
  • Ideal even for small-volume fermentors
  • Special internal pH electrode lets you calibrate without using gases
  • 1 year warranty
More About this Item
For use with Cole-Parmer® ion-selective meters.