Cole-Parmer Essentials Cheesecloth Wipes, 7" x 14"; 200/PK
Representative image only.
Cole-Parmer Essentials Item # WW-33670-15
Absorbs 6 times their own weight
  • Reagent-grade quality
  • Clean any optical-grade surface
  • 100% Cotton
  • 1 year warranty
PRODUCT OPTIONS
₹4,000.00INR / Pkg of 200
SPECS & DESCRIPTION
Specifications & Description
  • Width (in)7
  • Length (in)14
  • DescriptionCheesecloth Wipes, 7\" x 14\", 200/pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Reagent-grade quality
  • Clean any optical-grade surface
  • 100% Cotton
  • 1 year warranty
More About this Item
These cheesecloth wipes absorbs six times their own weight of both solvents and aqueous solutions. They meet all lab requirements and won't scratch surfaces or fall apart like paper towels.