Cole-Parmer CG-900 Cryo-Blade™ Cryogenic Processor Inner Bowl, 5 L
Representative image only.
Cole-Parmer Item # WW-61211-70
Optimize performance of your Cole-Parmer® Cryo-Blade™ Cryogenic Grinder
  • Accessories to rapidly pulverize samples from bulk to analytical preparation
  • Excellent for cryogenic grinding of fruits, vegetables, seeds, stalks, roots, fungi, and flowers
PRODUCT OPTIONS
₹33,500.00INR / Each
Cryo-Blade™ Cryogenic Mill
SPECS & DESCRIPTION
Specifications & Description
  • Jar Capacity (L)5
  • Compatible WithCryo-Blade™ Cryogenic Grinder 61211-71 and 61211-72
  • DescriptionCryo-Blade™ Cryogenic Processor Inner Bowl, 5 L
Key Features
  • Accessories to rapidly pulverize samples from bulk to analytical preparation
  • Excellent for cryogenic grinding of fruits, vegetables, seeds, stalks, roots, fungi, and flowers
More About this Item
Suitable for use with the Cole-Parmer® CG-900 Cryo-Blade™ cryogenic grinders, 61211-71 and 61211-72. Mills sold separately.
Removable 5 L inner bowl can accommodate samples up to 1 kg and allows samples to be stored in the freezer, giving the user the option of using a second bowl to prepare a new batch.