Cole-Parmer CDI-700 Monobed Deionizer, clear housing, 3/4" NPT or 3/8" hose barb
Representative image only.
Cole-Parmer Item # WW-01503-20
Reduce harmful ions in your system
  • Monitor water quality with color indicating resin
  • Purify water up to 15 MΩ-cm
  • 1 year warranty
₹25,100.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • ASTM Water GradeType I
  • Capacity1050 grains as CaCO3
  • Max Flow Rate (L/hour)57
  • Max Flow Rate (GPH)15
  • Inlet Port3/4\" NPT(F)
  • Max Inlet Water Temperature (° F)90
  • Max Inlet Water Temperature (° C)32
  • Max Inlet Pressure (PSI)125
  • Max Inlet Pressure (bar)8.6
  • DescriptionMonobed Deionizer, clear housing, 3/4\" NPT or 3/8\" hose barb
  • Warranty1 Year
1 Year Warranty
Key Features
  • Monitor water quality with color indicating resin
  • Purify water up to 15 MΩ-cm
  • 1 year warranty
More About this Item
The unique clear housing lets you see when the resin is exhausted. The deionizer is loaded with a mixed bed resin (1:1 equivalent) with a maximum capacity of 700 grains as CaCO3. The resin is specially treated to yield a gradual color change from dark blue to amber to indicate the level of exhaustion (spending). Change the resin when all but 1" (2.5 cm) of resin is amber. Replace exhausted resin quickly during a momentary shutdown of the water system. Order resin separately (deionizer requires up to two resin refills).