Cole-Parmer Casting Clamp for Dual Vertical Mini-Gel Systems
Representative image only.
Cole-Parmer Item # WW-28565-32
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCasting Clamp for Dual Vertical Mini-Gel Systems
  • Warranty3 year
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
Casting clamp for clamping the glass plate set during gel casting. Chemical- and corrosion-resistant polypropylene. Three needed per plate set for mini units; six needed per plate set for adjustable-height units