Cole-Parmer BioProducer 12 Reuse Hollow Fiber Filter, 1.25 m2, Feed: 1.5" TC; 1/Pk
Representative image only.
Cole-Parmer Item # WW-15959-95
Uniquely modified PES hollow fiber membranes for biopharmaceutical applications
  • • Open channel structure for cell and particle compatibility in the process stream
  • • Available with MWCO from 3K to 750K (5K, 10K, 30K, 50K, 100K, 300K, 500K and 750K) as well as 0.1 µm, 0.2 µm, and 0.45 µm
  • • Minimized batch-to-batch variation for consistent performance
  • • 100% integrity tested with COA with each finished product
  • • Complete validation of membrane processes and cartridge assemblies
  • • Applications include perfusion and upstream/downstream processes
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Connections3/8\" hose barb
  • Effective Filtration Area (cm²)2600
  • DescriptionReuse Hollow Fiber Filter, 1.25 m2, Feed: 1.5\" TC; 1/Pk
Key Features
  • • Open channel structure for cell and particle compatibility in the process stream
  • • Available with MWCO from 3K to 750K (5K, 10K, 30K, 50K, 100K, 300K, 500K and 750K) as well as 0.1 µm, 0.2 µm, and 0.45 µm
  • • Minimized batch-to-batch variation for consistent performance
  • • 100% integrity tested with COA with each finished product
  • • Complete validation of membrane processes and cartridge assemblies
  • • Applications include perfusion and upstream/downstream processes
More About this Item
“ReUse” Reusable Hollow Fiber Cartridges
Compatible with most automatic process development systems
Ideal for standard applications where repeated cleaning is needed
Can withstand CIP
MiniDiscover and Discover are small-volume hollow fiber cartridges for initial membrane applications with sample volumes between 10 to 400 mL. Compatible with most automated process development systems.
Explorer is developed for membrane screening, scale-down studies, and processing of smaller sample volumes between 150 mL and 3 L.
BioProducer is a large-scale high-capacity hollow fiber cartridge for product batches up to several thousand liters.
Maximizer is a group of hollow fiber cartridges for large-scale processing of biopharmaceuticals and food and beverage products up to 1000 L/cartridge.
“Green” Single-Use Hollow Fiber Cartridges
Antifouling ultrafiltration membranes for improved yields
Gamma irradiated, glycerine free, low binding
Lower extractable levels than a glycerine conditioned membrane
Ready to use without any prerinse or sanitization
DiscoverGreen are ¼” cartridges for small volume membrane screening and product discovery applications. Sample volumes between 10 to 400 mL.
ExplorerGreen are ½” cartridges for cross-flow development work and laboratory scale applications. Sample volumes between 10 to 1500 mL.