Cole-Parmer Antifoam Meter/Controller, (LED bar graph)
Representative image only.
Cole-Parmer Item # WW-01973-02
Controls foaming action via antifoam agent
  • Designed for panel mounting
  • Set relay to delay action from 0 to 5 seconds
  • 5 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialAnodized aluminum front panel, epoxy-coated metallic case
  • Height (in)3.5
  • Width (in)9.5
  • Depth (InchShort)6.875
  • Power (VAC)110 / 220
  • DescriptionAntifoam Meter/Controller, (LED bar graph)
  • Warranty5 Years
5 Year Warranty
Key Features
  • Designed for panel mounting
  • Set relay to delay action from 0 to 5 seconds
  • 5 year warranty
More About this Item
Prevents contamination and promotes optimal growth in your microbial fermentation system. LED bar graph indicates when foam/media level exceeds your set point; meter then activates a relay for a pump, valve, or alarm.