Cole-Parmer All-in-One pH/ATC Probe, Refillable/DJ/Glass
Representative image only.
Cole-Parmer Item # WW-05991-36
Use these electrodes with most Oakton handheld or benchtop pH meters for accurate and reliable readings
  • Refillable electrode provides more accurate pH measurement
  • Amber glass bulb minimizes sodium error in high pH solutions
  • Use at temperatures up 110°C
  • 1 year warranty
₹33,100.00INR / Each
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsRefillable
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Min pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)110
  • Temperature CompensationYes
  • ATC ConnectorAudio plug, 2.5 mm
  • ATC ElementThermistor
  • ConnectorBNC
  • Cable Length (m)1
  • DescriptionAll-in-One pH/ATC Probe, Refillable/DJ/Glass
  • Warranty1 year
1 Year Warranty
Key Features
  • Refillable electrode provides more accurate pH measurement
  • Amber glass bulb minimizes sodium error in high pH solutions
  • Use at temperatures up 110°C
  • 1 year warranty
More About this Item
These glass 'All-in-one' pH/temperature electrodes offer more reliable and stable reading than epoxy versions. Amber glass bulb for low sodium error even in solutions with a pH greater than 12. Double-junction design assures longer life even with dirty or troublesome samples such as TRIS buffer or wastewater. A 2.5 mm mini audio plug combined with a thermistor for automatic temperature compensation input to your Oakton pH meter. Compatible with the following Oakton meters: handheld pH/Ion 5, 6, 11, 110 and benchtop 510, 700, 1100, 2100, and 2700.