This item is now discontinued. We recommend the following as suggested replacements: WW-24502-18
Cole-Parmer A23145PRX Filter Pipette Tips, sterile, 2" L, 10
Representative image only.
Cole-Parmer Item # WW-25710-18
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • AutoclavableYes
  • Max Volume (µL)100
  • Max Volume (mL)0.1
  • Overall Length (in)2
  • Overall Length (mm)50.8
  • Color CodeNatural
  • Compatible withCostar® Octapette™; Eppendorf® 2000, 2100, 4703, 4810; Finnipipette® digital, Techpette® fixed/variable; MLA®; Oxford® 3000, BenchMate™, -SV, multi-channel; Pipetman® P-20; Wheaton Micropettors
  • DescriptionFilter Pipette Tips, sterile, 2\" L, 10
More About this Item
These state-of-the-art polypropylene pipette tips feature a unique, hydrophobic super filter. The inert filter prevents liquid bypass in the event of over-pipetting and doesn't seal or trap liquid—allowing for non-contaminated sample recovery. Filters also eliminate trace carryover and contamination by aerosols generated during sample aspiration.
Autoclavable tips are available either non-sterile or gamma-irradiation sterilized, and packaged in racks of 96 or 100. Each rack is stackable, shrink-wrapped with a tear-off seal, and labeled with pipette type and lot number.