Cole-Parmer 410-0001-FLS-CP Cell Strainer, Blue, 40 um
Representative image only.
Cole-Parmer Item # WW-06059-85
Obtain clump and debris-free, single-cell suspensions
  • Fit right over your 50-mL centrifuge tube
  • Choose from 40, 70 or 100 µm mesh sizes
  • Easily identify pore size by support color
  • Gamma sterilized and individually wrapped
SPECS & DESCRIPTION
Specifications & Description
  • Pore Size (micron)40
  • Housing MaterialPolypropylene
  • SterileYes
  • DescriptionCell Strainer, Blue, 40 um; 50/Cs
Sterile
Key Features
  • Fit right over your 50-mL centrifuge tube
  • Choose from 40, 70 or 100 µm mesh sizes
  • Easily identify pore size by support color
  • Gamma sterilized and individually wrapped
More About this Item
Obtain uniform cell suspensions with these sterile cell strainers. Housing features extended tab for easy handling.