Cole-Parmer 2251S Vial Cap for 15 mL Short Vial, Polypropylene with Silicone Liner; 100/Pk
Representative image only.
Cole-Parmer Item # WW-41071-27
Safely grind samples in the HG-600 Geno/Grinder® 2010 and HG-400 MiniG® homogenizers
  • Screw thread cap ensures a tight seal
  • Constructed with a polypropylene cap and silicone liner
  • Compatible with 04576-99 and 04575-71, 15 mL polycarbonate vials
PRODUCT OPTIONS
₹30,600.00INR / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene Cap with Silicone Liner
  • ColorWhite
  • Compatible With04576-99 and 04575-71 Vials
  • ConnectionsThread
  • DescriptionVial Cap for 15 mL Short Vial, Polypropylene with Silicone Liner; 100/Pk
Key Features
  • Screw thread cap ensures a tight seal
  • Constructed with a polypropylene cap and silicone liner
  • Compatible with 04576-99 and 04575-71, 15 mL polycarbonate vials
More About this Item
These polypropylene caps are lined with a thick 3/32” red silicone disc that prevents cracking. Threaded caps firmly seal Cole-Parmer® homogenizer vials. Replacement caps for 04576-99 and 04575-71. Vials sold separately.
These polypropylene caps are lined with a thick 3/32” red silicone disc that prevents cracking.