Cole-Parmer 1261AG4H30 Propane primary regulator, 7.5 cu ft disposable cylinder
Representative image only.
Cole-Parmer Item # WW-02655-30
Provides controlled presssure of gas
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPropane primary regulator, 7.5 cu ft disposable cylinder
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
A primary regulator is required to adjust the flow rate of fuel gas entering the flame photometer. Choose a regulator from the table according to your fuel type and cylinder valve outlet.
Note: All fuels should be free of heavy hydrocarbon deposits and regulated at the cylinder to 30 psi.