Cole-Parmer Essentials 100 Sterile Vials with caps in foam rack, 2 mL
Representative image only.
Cole-Parmer Essentials Item # WW-36270-70
Protect your samples
  • Constructed of durable polypropylene
  • Secure your sample with the O-ring lined cap
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)2
  • SterileYes
  • Description100 Sterile Vials with caps in foam rack, 2 mL
  • Warranty1 Year
1 Year Warranty
Key Features
  • Constructed of durable polypropylene
  • Secure your sample with the O-ring lined cap
  • 1 year warranty
More About this Item
These 2-mL vials are ideal for use with the Mini-BeadBeater™ Mill 36270-02 and -07. These vials have an ID of 0.9 mm with an OD of 1.1 mm. The vial length without lid is 4.5 mm and with lid is 4.7 mm.