Cole-Parmer 0.990/1.040kg/L Draft Survey Hydrometer
Representative image only.
Cole-Parmer Item # WW-90310-51
Easily verify apparent density of sea/fresh water
  • Temperature standard is 60⁰F (15.6⁰C)
  • Individually serialized for easy identification
  • Ensure accuracy with multi-language statement of accuracy supplied by manufacturer
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")13
  • Hydrometer FormPlain
  • Alcohol Range (Min) (%)0
  • Division0.0005 kg/L
  • Accuracy±0.0005 kg/L
  • Length (mm)330
  • Description 0.990/1.040 kg/L Draft Survey Hydrometer
  • Warranty3 month
3 Month Warranty
Key Features
  • Temperature standard is 60⁰F (15.6⁰C)
  • Individually serialized for easy identification
  • Ensure accuracy with multi-language statement of accuracy supplied by manufacturer
  • 3 month warranty
More About this Item
The draft survey hydrometer, with a range from 0.990 to 1.040 kg/L, easily measures the apparent density of sea/fresh water. These measurements help determine the amount of cargo that can be loaded onto bulk carrying cargo vessels for the fishing and shipping industries. The glass hydrometer has 0.0005 kg/L divisions and a lead free ballast.
Equivalent to H-B Instrument 618090500.