Clean Aire II Ductless Fume Hood, 30" W; 230 VAC

Representative image only.
Mfr # 93025-2 – Item # WW-33706-25
No need for expensive and disruptive duct installation
  • Activated carbon filter removes fumes, odors, and nonhazardous chemicals
  • Fire retardant fiberglass construction is durable and safe
  • Ships completely assembled and ready for operation
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Hood Width (in)30
  • Blower TypeIntegral
  • LightsVapor-proof
  • Exterior Width (in)30
  • Exterior Height (in)36
  • Exterior Depth (in)23
  • Interior Width (in)24
  • Interior Height (in)18.5
  • Interior Depth (in)21
  • Power (VAC)230
  • Power (Hz)50
  • Sash opening (in)10
  • Exhaust Volume (CFM)274
  • Shipping Weight (lbs)150
  • Shipping Weight (kg)68
  • Filter TypeActivated carbon
  • Number of Filters Required1
  • DescriptionDuctless Fume Hood
  • Warranty1 Year
1 Year WarrantyISO9001 Certified SupplierMade in the U.S.A.

Key Features

  • Activated carbon filter removes fumes, odors, and nonhazardous chemicals
  • Fire retardant fiberglass construction is durable and safe
  • Ships completely assembled and ready for operation
  • 1 year warranty

More About this Item

These ductless fume hoods feature molded fiberglass construction for self-containment and safety. Each hood is equipped with vapor-proof lights, integral carbon filter and internal blower. Small amounts of solvents, acids, and nuisance odors are adsorbed by the carbon and the cleaned air returned to the room. Installation does not require access to ventilation ducts and saves on energy, since a large volume external blower is not necessary. Order work surface and base separately.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat