Clavies Biohazard Bag Stand with Tray for 15 to 20 gal Bags; 1/Pk
Representative image only.
Item # WW-06497-18
Securely supports biohazard bags and contents on benchtop or floor
  • Stands ship flat and are easy to assemble
  • Polypropylene bottom serves as containment tray
PRODUCT OPTIONS
₹16,500.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (Gal)20
  • ColorOrange and silver
  • Compatible WithBiohazard Bags
  • DescriptionF13192-0003 Clavies Biohazard Bag Stand with Tray for 15 to 20 gal Bags; 1/Pk
Key Features
  • Stands ship flat and are easy to assemble
  • Polypropylene bottom serves as containment tray
More About this Item
Clavies stainless steel and polypropylene bag stands are designed to securely support biohazard bags and their contents. Tray is safety orange for easy identification as a biohazard containment system. Stands are lightweight and easy to assemble. Autoclavable
Fits bags: 24 x 36" (61 x 91 cm)