CINC 02W1.075B-2 Centrifugal Multi-Purpose Separator/Extractor Weir; 1.075" Diameter
Representative image only.
CINC Mfr # 02W1.075B-2 – Item # WW-17315-20
Mix, react, and/or separate liquid-liquid mixtures in a single device
  • Highly flexible design operates effectively under changing inlet concentrations, variable flow rates, and flow disruptions
  • Interchangeable weirs allow for broad applications in research, pilot plant, or production operations
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCentrifugal Multi-Purpose Separator/Extractor Weir; 1.075" Diameter
Key Features
  • Highly flexible design operates effectively under changing inlet concentrations, variable flow rates, and flow disruptions
  • Interchangeable weirs allow for broad applications in research, pilot plant, or production operations
More About this Item
Weirs are available in various diameters. Use the large diameter weirs in applications with high-viscosity liquids.