Chromatography Research Supplies Puresep-P 230660 Puresep™ P Septa, 1/2" (12.5 mm); 50/Pk
Representative image only.
Chromatography Research Supplies Mfr # 230660 – Item # WW-01994-03
Designed for enhanced bond and durability
  • Constructed of silicone rubber with a stable layer of polyimide film
SPECS & DESCRIPTION
Specifications & Description
  • Diameter (in)0.5
  • DescriptionSepta, 1/2\" (12.5 mm); 50/pack
Key Features
  • Constructed of silicone rubber with a stable layer of polyimide film
More About this Item
PURESEP-P™ septa are made of silicone rubber with a stable layer of polyimide film; withstand temperatures to 572°F (300°C).