Chill packs for waterless ice bucket
Representative image only.
Lab Armor Item # WW-06274-29
Replace water in laboratory water baths and ice in ice buckets.
  • Reduce the risk of contamination, achieve reproducible results and have greater confidence in your work
  • No need to worry about burnout as there is no water to evaporate.
  • Highly recyclable, fewer harmful biocides, and more energy efficient beads are eco-friendly
  • Reliably cold chill packs
  • 1 year warranty
PRODUCT OPTIONS
₹7,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionChill packs for waterless ice bucket
  • Warranty1 year
1 Year Warranty
Key Features
  • Reduce the risk of contamination, achieve reproducible results and have greater confidence in your work
  • No need to worry about burnout as there is no water to evaporate.
  • Highly recyclable, fewer harmful biocides, and more energy efficient beads are eco-friendly
  • Reliably cold chill packs
  • 1 year warranty
More About this Item
Compatible with waterless ice buckets, items 06274-24 and -28.
Includes four chill packs for use with the Waterless Ice Buckets.
These hold temperature for a long time and they are the tough enough to be used over and over again without breaking, or tearing like the vinyl shipping freeze packs will. And after a lot of use, they can be cleaned with alcohol or bleach to keep the Chill Bucket system free of mold and other microbes.