Chemware D1069076 Erlenmeyer Perfluoroalkoxy (PFA) Flask, 200 mL, 1/Pk
Representative image only.
Chemware Mfr # D1069076 – Item # WW-06112-05
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPerfluoroalkoxy (PFA)
  • Capacity (mL)200
  • Stopper Number Accepted4
  • Overall Height (cm)12.3
  • DescriptionErlenmeyer Perfluoroalkoxy (PFA) Flask, 200 mL, 1/Pk
More About this Item
Flasks are ideal for ultrapure applications. They can be heated in microwave; maximum temperature is 500degF (260degC). Graduated, chemically inert flasks are transparent for easy viewing of contents.