Chem-Tainer Conical-Bottom Tank System, LDPE, 200 gallon
Representative image only.
Chem-Tainer Mfr # TC3165CA-200GAL FA0150MO-TC3165AF-TC3165CM – Item # WW-06925-20
Systems include tank, stand, cover, and bulkhead fittings
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyethylene
  • Volume (Gal)200
  • Volume (Liters)757
  • Height (in)66 7/8
  • Height (cm)169.8625
  • Max Temperature (° F)150
  • Max Temperature (° C)65
  • DescriptionConical-Bottom Tank System, LDPE, 200 gallon
More About this Item
Linear polyethelyne tanks have conical bottoms for complete drainage of fluids and are impact and chemical resistant. Tanks can withstand continuous operating temperature of 150°>F (65°>C). Translucent sides let you observe liquid levels.
Complete tank systems include tank, carbon steel stand with agitator mounting, LDPE cover, and bulkhead fitting—indicate fitting placement at time of order. Order replacement tanks separately.