Chem-Tainer Conical-Bottom Open-Top Tank with Stand, PP, 200 gallon
Representative image only.
Chem-Tainer Mfr # TC3175BP-FA0150-INSTALLED – Item # WW-06929-16
Tanks give you excellent chemical compatibility
PRODUCT OPTIONS
₹661,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Volume (Gal)200
  • Volume (Liters)757
  • Height (in)75
  • Height (cm)190.5
  • Outside Diameter (OD) (in)31
  • Max Temperature (° F)500
  • Max Temperature (° C)260
  • DescriptionConical-Bottom Open-Top Tank with Stand, PP, 200 gallon
More About this Item
Tanks are available in polypropylene or polyethylene; all stands are made from one-piece molded polyethylene. There are no metal parts—eliminate rust and corrosion problems.