CELLTREAT Scientific Products 229875 Vented Sterile Fernbach Flasks with Baffled Bottom, 3L, Individually Wrapped; 6/cs
Representative image only.
CELLTREAT Scientific Products Mfr # 229875 – Item # WW-01931-81
Get improved gas exchange via oversized vent area
  • Fernbach style offers a large surface area to volume ratio
  • Fernbach flasks with baffled bottom
  • Fernbach flask shape increases surface area to volume ratio
  • Baffled bottoms aid agitation and aeration during shaking
PRODUCT OPTIONS
₹61,300.00INR / Case of 6
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolycarbonate
  • Capacity (mL)4000
  • AutoclavableYes
  • Working Volume (mL)3000
  • DescriptionVented Sterile Fernbach Flasks with Baffled Bottom, 3L, Individually Wrapped; 6/cs
Sterile
Key Features
  • Fernbach style offers a large surface area to volume ratio
  • Fernbach flasks with baffled bottom
  • Fernbach flask shape increases surface area to volume ratio
  • Baffled bottoms aid agitation and aeration during shaking
More About this Item
Designed for suspension culture applications, these flasks feature 70-mm opening for easy filling and dispensing. Constructed of optically clear, autoclavable polycarbonate. All flasks are supplied sterile (gamma irradiated) and individually double bagged.