CELLTREAT Scientific Products 229695 Sterile Petri Dishes, 100 x 15 mm; 500/cs
Representative image only.
CELLTREAT Scientific Products Mfr # 229695 – Item # WW-01932-29
Optically clear for improved imaging and observation
  • Consistent flatness allows for even media distribution
  • Manufactured from virgin polystyrene
  • All dishes are supplied sterile (gamma irradiated)
  • 15 to 16 mL working volume
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSterile Petri Dishes, 100 x 15 mm; 500/cs
Sterile
Key Features
  • Consistent flatness allows for even media distribution
  • Manufactured from virgin polystyrene
  • All dishes are supplied sterile (gamma irradiated)
  • 15 to 16 mL working volume
More About this Item
Flared lid skirt and squared corners provide easy one-handed operation, even while wearing gloves. Lids have molded spacers to reduce condensation and allow air circulation. Stacking rings on the lids provide increased stability.
All dishes are supplied sterile (gamma irradiated) and come packaged 25 dishes/bag.