CE International Trading Mesh Screen, SS, 2.8 mm Sieve Opening, 7 mesh, 18" dia
Representative image only.
CE International Trading Mfr # LS450S005-#7MESH – Item # WW-59979-43
Mesh screens separate oversized particles from your sample
PRODUCT OPTIONS
₹38,800.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless Steel
  • U.S. Standard Screen Size (No.)7
  • Tyler Standard Screen Size (mesh)7
  • Sieve Opening (mm)2.83
  • Outside Diameter (in)18
  • DescriptionMesh Screen, SS, 2.8 mm sieve opening, 7 mesh
More About this Item
Screens are 18" (45.7 cm) in diameter with holes for securing in the screen assembly (sold separately under part number 59998-54).