CE International Trading Mesh Screen, Nylon, 4.00 mm Sieve Opening, 5 mesh, 18" dia
Representative image only.
CE International Trading Mfr # LS450N006-#5MESH – Item # WW-59979-70
Mesh screens separate oversized particles from your sample
PRODUCT OPTIONS
₹38,500.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNylon
  • U.S. Standard Screen Size (No.)5
  • Tyler Standard Screen Size (mesh)5
  • Sieve Opening (mm)4
  • Outside Diameter (in)18
  • DescriptionMesh Screen, Nylon, 4.00 mm sieve opening, 5 mesh
More About this Item
Screens are 18" (45.7 cm) in diameter with holes for securing in the screen assembly (sold separately under part number 59998-54).